Carvon
Lieferant: EHRENSTORFER
Synonyme:
2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone
C11052000
EHERC11052000EA
57
EUR
EHERC11052000
Carvon
Carvon
Formel:
C₁₀H₁₄O Molecular Weight: 150.2176 g/mol |
MDL:
MFCD00062996 CAS-Nummer: 99-49-0 EINECS: 202-759-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...