D(+)-Cellobiose
Lieferant: EHRENSTORFER
Synonyme:
β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
C11067000
EHERC11067000EA
48
EUR
EHERC11067000
D(+)-Cellobiose
D(+)-Cellobiose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342,30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136034 CAS-Nummer: 528-50-7 EINECS: 208-436-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...