cis-Chlordan

Lieferant: EHRENSTORFER

C11201000
EHERC11201000EA 102 EUR
EHERC11201000
cis-Chlordan
cis-Chlordane
Formel: C₁₀H₆Cl₈
CAS-Nummer: 5103-71-9
EINECS: 225-825-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR