trans-Chlordan

Lieferant: EHRENSTORFER

C11202000
EHERC11202000EA 102 EUR
EHERC11202000
trans-Chlordan
trans-Chlordane
Formel: C₁₀H₆Cl₈
CAS-Nummer: 5103-74-2
EINECS: 225-826-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR