Chlordimeform Hydrochlorid

Lieferant: EHRENSTORFER

C11240100
EHERC11240100EA 44 EUR
EHERC11240100
Chlordimeform Hydrochlorid
Chlordimeform Hydrochlorid
Formel: C₁₀H₁₄Cl₂N₂
Molecular Weight: 233.14 g/mol
CAS-Nummer: 19750-95-9
EINECS: 243-269-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR