Chlorfenethol
Lieferant: EHRENSTORFER
Synonyme:
1,1-Bis(4-chlorophenyl)ethanol, 4,4'-Dichloro-alpha-methylbenzhydrol
C11250000
EHERC11250000EA
78
EUR
EHERC11250000
Chlorfenethol
Chlorfenethol
Formel:
C₁₄H₁₂Cl₂O |
MDL:
MFCD00000628 CAS-Nummer: 80-06-8 EINECS: 201-246-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...