Chlorflurenol-Methylester (2-Chlor-9-hydroxy-9H-fluoren-9-carbonsäuremethylester)

Lieferant: EHRENSTORFER

C11305000
EHERC11305000EA 90 EUR
EHERC11305000
Chlorflurenol-Methylester (2-Chlor-9-hydroxy-9H-fluoren-9-carbonsäuremethylester)
Chlorflurenol-Methylester (2-Chlor-9-hydroxy-9H-fluoren-9-carbonsäuremethylester)
Formel: C₁₅H₁₁ClO₃
CAS-Nummer: 2536-31-4
EINECS: 219-800-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR