Chlorhexidin
Lieferant: EHRENSTORFER
Synonyme:
1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]
C11310000
EHERC11310000EA
72
EUR
EHERC11310000
Chlorhexidin
Chlorhexidin
Formel:
C₂₂H₃₀Cl₂N₁₀ Molecular Weight: 505,4466 g/mol |
MDL:
MFCD00009673 CAS-Nummer: 55-56-1 EINECS: 200-238-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...