2-Chlor-N-(2,6-diethylphenyl)acetamid
Lieferant: EHRENSTORFER
Synonyme:
2-Chloro-2',6'-diethylacetanilide
C11349700
EHERC11349700EA
134
EUR
EHERC11349700
2-Chlor-N-(2,6-diethylphenyl)acetamid
2-Chlor-N-(2,6-diethylphenyl)acetamid
Formel:
C₁₂H₁₆ClNO Molecular Weight: 225,72 g/mol |
MDL:
MFCD00124800 CAS-Nummer: 6967-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...