2-Chlor-4,6-dinitroanilin

Lieferant: EHRENSTORFER

Synonyme: 6-Chlor-2,4-dinitroanilin

C11405200
EHERC11405200EA 41 EUR
EHERC11405200
2-Chlor-4,6-dinitroanilin
2-Chlor-4,6-dinitroanilin
Formel: C₆H₄ClN₃O₄
Molecular Weight: 217,57 g/mol
Schmelzpunkt: 248 °C
MDL: MFCD00007147
CAS-Nummer: 3531-19-9
EINECS: 222-564-9
UN: 2237
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR