2-Chlor-4-(methylsulfonyl)benzoesäure

Lieferant: EHRENSTORFER

Synonyme: 2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid

C11447000
EHERC11447000EA 97 EUR
EHERC11447000
2-Chlor-4-(methylsulfonyl)benzoesäure
2-Chloro-4-(methylsulphonyl)benzoic acid
Formel: C₈H₇ClO₄S
Molecular Weight: 234,66 g/mol
MDL: MFCD00216496
CAS-Nummer: 53250-83-2
EINECS: 406-520-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR