2-Chlor-4-nitrophenol
Lieferant: EHRENSTORFER
Synonyme:
1-Chloro-2-hydroxy-5-nitrobenzene
C11454000
EHERC11454000EA
53
EUR
EHERC11454000
2-Chlor-4-nitrophenol
2-Chlor-4-nitrophenol
Formel:
C₆H₄ClNO₃ Molecular Weight: 173,56 g/mol Schmelzpunkt: 109…112 °C |
MDL:
MFCD00043910 CAS-Nummer: 619-08-9 EINECS: 210-578-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...