2-Chlorphenol-[Ring-D4]

Lieferant: EHRENSTORFER
Achtung

Synonyme: 2-Chlorphenol (Ring-D4), 1-Chlor-2-hydroxybenzol-D4, 2-Chlor-1-hydroxybenzol-D4

C11470100
EHERC11470100EA 186 EUR
EHERC11470100
2-Chlorphenol-[Ring-D4]
2-Chlorphenol-[Ring-D4]
Formel: C₆HD₄ClO
Molecular Weight: 132,53 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 93951-73-6
UN: 2021
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR