4-Chlorphenylacetat

Lieferant: EHRENSTORFER

C11473200
EHERC11473200EA 87 EUR
EHERC11473200
4-Chlorphenylacetat
4-Chlorphenylacetat
Formel: C₈H₇ClO₂
Molecular Weight: 170,5930 g/mol
MDL: MFCD00623648
CAS-Nummer: 876-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR