(3-Chlorphenoxy)essigsäure
Lieferant: EHRENSTORFER
Synonyme:
3-CPA
C11479500
EHERC11479500EA
52
EUR
EHERC11479500
(3-Chlorphenoxy)essigsäure
(3-Chlorphenoxy)essigsäure
Formel:
(C₆H₄Cl)OCH₂COOH Molecular Weight: 186,59 g/mol Siedepunkt: 308 °C (1013 hPa) Schmelzpunkt: 110 °C |
MDL:
MFCD00015850 CAS-Nummer: 588-32-9 EINECS: 209-616-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...