2-(4-Chlorphenoxy)isobuttersäure

Lieferant: EHRENSTORFER

Synonyme: 2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid

C11484000
EHERC11484000EA 50 EUR
EHERC11484000
2-(4-Chlorphenoxy)isobuttersäure
2-(4-Chlorphenoxy)isobuttersäure
Formel: C₁₀H₁₁ClO₃
Molecular Weight: 214,65 g/mol
MDL: MFCD00004192
CAS-Nummer: 882-09-7
EINECS: 212-925-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR