4-Chlorphenylharnstoff
Lieferant: EHRENSTORFER
Synonyme:
1-(4-Chlorophenyl)urea
C11492000
EHERC11492000EA
61
EUR
EHERC11492000
4-Chlorphenylharnstoff
4-Chlorphenylharnstoff
Formel:
C₇H₇ClN₂O Molecular Weight: 170,6 g/mol |
MDL:
MFCD00014788 CAS-Nummer: 140-38-5 EINECS: 205-412-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...