5,7-Dichlor-2-methyl-8-chinolinol
Lieferant: EHRENSTORFER
Synonyme:
5,7-Dichloro-8-hydroxy-2-methylquinoline, 5,7-Dichloro-2-methylquinolin-8-ol
C11505800
EHERC11505800EA
81
EUR
EHERC11505800
5,7-Dichlor-2-methyl-8-chinolinol
5,7-Dichlor-2-methyl-8-chinolinol
Formel:
C₁₀H₇Cl₂NO Molecular Weight: 228,0747 g/mol |
MDL:
MFCD00023984 CAS-Nummer: 72-80-0 EINECS: 200-789-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...