Chlorphenesin

Lieferant: EHRENSTORFER

C11553000
EHERC11553000EA 66 EUR
EHERC11553000
Chlorphenesin
Chlorphenesin
Formel: C₉H₁₁ClO₃
Molecular Weight: 202.64 g/mol
Schmelzpunkt: 80
MDL: MFCD00021990
CAS-Nummer: 104-29-0
EINECS: 203-192-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR