Chlorpyrifos-methyl

Lieferant: EHRENSTORFER
Achtung

Synonyme: O,O-Dimethyl-O-3,5,6-trichlor-2-pyridylphosphorothionat, O,O-Dimethyl-O-3,5,6-trichlor-2-pyridylthiophosphat

C11601000
EHERC11601000EA 85 EUR
EHERC11601000
Chlorpyrifos-methyl
Chlorpyrifos-methyl
Formel: C₇H₇Cl₃NO₃PS
Molecular Weight: 322,54 g/mol
Schmelzpunkt: 45,5…46.5 °C
CAS-Nummer: 5598-13-0
EINECS: 227-011-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR