2,6-Dichlorthiobenzamid

Lieferant: EHRENSTORFER

Synonyme: Chlorthiamid, 2,6-Dichlorthiobenzoesäureamid

C11630000
EHERC11630000EA 95 EUR
EHERC11630000
2,6-Dichlorthiobenzamid
2,6-Dichlorthiobenzamid
Formel: C₇H₅Cl₂NS
Molecular Weight: 206,09 g/mol
MDL: MFCD00053523
CAS-Nummer: 1918-13-4
EINECS: 217-637-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR