4-Cholesten-3-on

Lieferant: EHRENSTORFER

Synonyme: (8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one, (+)-4-Cholesten-3-one

C11665300
EHERC11665300EA 91 EUR
EHERC11665300
4-Cholesten-3-on
4-Cholesten-3-one
Formel: C₂₇H₄₄O
Molecular Weight: 384,65 g/mol
Schmelzpunkt: 81…82 °C
MDL: MFCD00003663
CAS-Nummer: 601-57-0
EINECS: 210-005-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR