2-(3-Chlorphenoxy)propionsäure

Lieferant: EHRENSTORFER

C11688000
EHERC11688000EA 84 EUR
EHERC11688000
2-(3-Chlorphenoxy)propionsäure
2-(3-Chlorphenoxy)propionsäure
Formel: C₉H₉ClO₃
MDL: MFCD00009723
CAS-Nummer: 101-10-0
EINECS: 202-915-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR