Kupfer(II)hydroxid
Lieferant: EHRENSTORFER
Synonyme:
Kupferdihydroxid
C11698000
EHERC11698000EA
58
EUR
EHERC11698000
Kupfer(II)hydroxid
Copper(II) hydroxide
Formel:
H₂CuO₂ Molecular Weight: 97,56 g/mol Dichte: 3,37 g/cm³ (20 °C) |
MDL:
MFCD00010968 CAS-Nummer: 20427-59-2 EINECS: 243-815-9 UN: 3288 ADR: 6.1,III Merck Index: 12,02709 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...