Crimidin (2-Chlor-6-methylpyrimidin-4-yldimethylamin)

Lieferant: EHRENSTORFER

C11750000
EHERC11750000EA 66 EUR
EHERC11750000
Crimidin (2-Chlor-6-methylpyrimidin-4-yldimethylamin)
Crimidin (2-Chlor-6-methylpyrimidin-4-yldimethylamin)
Formel: C₇H₁₀ClN₃
CAS-Nummer: 535-89-7
EINECS: 208-622-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR