Cycloheximid
Lieferant: EHRENSTORFER
|
Gefahr
|
C11830000
EHERC11830000EA
66
EUR
EHERC11830000
Cycloheximid
Formel:
C₁₅H₂₃NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082346 CAS-Nummer: 66-81-9 EINECS: 200-636-0 UN: 2811 ADR: 6.1,I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...