DL-Cystin
Lieferant: EHRENSTORFER
Synonyme:
(H-DL-Cys-OH)2
C11925000
EHERC11925000EA
36
EUR
EHERC11925000
DL-Cystin
DL-Cystin
Formel:
C₆H₁₂N₂O₄S₂ Siedepunkt: 429 °C (1013 hPa) Schmelzpunkt: 227 °C |
MDL:
MFCD00084652 CAS-Nummer: 923-32-0 EINECS: 213-094-5 Merck Index: 13,02811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...