(2,3-Dichlorphenoxy)essigsäure
Lieferant: EHRENSTORFER
Synonyme:
2,3-D
C11939000
EHERC11939000EA
186
EUR
EHERC11939000
(2,3-Dichlorphenoxy)essigsäure
(2,3-Dichlorphenoxy)essigsäure
Formel:
(C₆H₃Cl₂)OCH₂COOH Molecular Weight: 221.04 g/mol Siedepunkt: 329 Schmelzpunkt: 195…198 °C |
MDL:
MFCD00004299 CAS-Nummer: 2976-74-1 EINECS: 221-022-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...