Daminozid (Bernsteinsäuremono(2,2-dimethylhydrazid))
Lieferant: EHRENSTORFER
Synonyme:
Daminozide, Succinic acid 2,2-Dimethylhydrazide
C11960000
EHERC11960000EA
54
EUR
EHERC11960000
Daminozid (Bernsteinsäuremono(2,2-dimethylhydrazid))
Daminozid (Bernsteinsäuremono(2,2-dimethylhydrazid))
Formel:
C₆H₁₂N₂O₃ |
MDL:
MFCD00002787 CAS-Nummer: 1596-84-5 EINECS: 216-485-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...