Dapson

Lieferant: EHRENSTORFER
Achtung

Synonyme: DDS, 4-Aminophenylsulfon, Bis(4-aminophenyl)sulfon, 4,4'-Sulfonyldianilin, 4,4'-Diaminodiphenylsulfon

C11963000
EHERC11963000EA 48 EUR
EHERC11963000
Dapson
Dapson
Formel: C₁₂H₁₂N₂O₂S
Molecular Weight: 248,31 g/mol
MDL: MFCD00007887
CAS-Nummer: 80-08-0
EINECS: 201-248-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR