4-(2,4-Dichlorphenoxy)buttersäure (2,4-DB)

Lieferant: EHRENSTORFER

C11980000
EHERC11980000EA 62 EUR
EHERC11980000
4-(2,4-Dichlorphenoxy)buttersäure (2,4-DB)
4-(2,4-Dichlorphenoxy)buttersäure (2,4-DB)
Formel: C₁₀H₁₀Cl₂O₃
MDL: MFCD00002819
CAS-Nummer: 94-82-6
EINECS: 202-366-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR