4-(2,4-Dichlorphenoxy)buttersäure(2-ethylhexyl)ester (2,4-DB-2-Ethylhexylester)
Lieferant: EHRENSTORFER
C11981000
EHERC11981000EA
71
EUR
EHERC11981000
4-(2,4-Dichlorphenoxy)buttersäure(2-ethylhexyl)ester (2,4-DB-2-Ethylhexylester)
4-(2,4-Dichlorphenoxy)buttersäure(2-ethylhexyl)ester (2,4-DB-2-Ethylhexylester)
Formel:
C₁₈H₂₆Cl₂O₃ |
CAS-Nummer:
1320-15-6 EINECS: 215-297-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...