4-(2,4-Dichlorphenoxy)buttersäuremethylester (2,4-DB-Methylester)
Lieferant: EHRENSTORFER
Synonyme:
Methyl 4-(2,4-dichlorophenoxy)butanoate
C11982000
EHERC11982000EA
80
EUR
EHERC11982000
4-(2,4-Dichlorphenoxy)buttersäuremethylester (2,4-DB-Methylester)
4-(2,4-Dichlorphenoxy)buttersäuremethylester (2,4-DB-Methylester)
Formel:
C₁₁H₁₂Cl₂O₃ |
CAS-Nummer:
18625-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...