2,2-Bis(4-chlorphenyl)ethanol

Lieferant: EHRENSTORFER

Synonyme: 4,4'-DDOH

C12070000
EHERC12070000EA 57 EUR
EHERC12070000
2,2-Bis(4-chlorphenyl)ethanol
2,2-Bis(4-chlorphenyl)ethanol
Formel: C₁₄H₁₂Cl₂O
Molecular Weight: 267,15 g/mol
CAS-Nummer: 2642-82-2
EINECS: 220-148-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR