Dexamethason-21-acetat
Lieferant: EHRENSTORFER
Synonyme:
11β,17α-Dihydroxy-21-(ethanoyloxy)-9-fluor-16α-methyl-1,4-pregnadien-3,20-dion, Dexamethasone acetate
C12170410
EHERC12170410EA
86
EUR
EHERC12170410
Dexamethason-21-acetat
Dexamethason-21-acetat
Formel:
C₂₄H₃₁FO₆ Molecular Weight: 434,5 g/mol |
MDL:
MFCD00027407 CAS-Nummer: 1177-87-3 EINECS: 214-646-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...