2,4-Diaminoanisol Dihydrochlorid

Lieferant: EHRENSTORFER

Synonyme: 4-Methoxybenzene-1,3-diamine dihydrochloride

C12192000
EHERC12192000EA 52 EUR
EHERC12192000
2,4-Diaminoanisol Dihydrochlorid
2,4-Diaminoanisol Dihydrochlorid
Formel: C₇H₁₂Cl₂N₂O
MDL: MFCD00054337
CAS-Nummer: 614-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR