4-Chlor-2,6-diaminopyrimidin

Lieferant: EHRENSTORFER

Synonyme: 2,4-Diamino-6-chloropyrimidine, 6-Chloropyrimidine-2,4-diamine

C12193500
EHERC12193500EA 79 EUR
EHERC12193500
4-Chlor-2,6-diaminopyrimidin
4-Chloro-2,6-diaminopyrimidine
Formel: C₄H₅ClN₄
Molecular Weight: 144,56 g/mol
MDL: MFCD00006097
CAS-Nummer: 156-83-2
EINECS: 205-863-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR