4,4'-Methylendi-o-toluidin

Lieferant: EHRENSTORFER

Synonyme: 4,4'-Methylenebis(2-methylaniline)

C12194800
EHERC12194800EA 83 EUR
EHERC12194800
4,4'-Methylendi-o-toluidin
4,4'-Methylendi-o-toluidin
Formel: C₁₅H₁₈N₂
Molecular Weight: 226,3200 g/mol
MDL: MFCD00126963
CAS-Nummer: 838-88-0
EINECS: 212-658-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR