2-Nitro-p-phenylendiamin
Lieferant: EHRENSTORFER
Synonyme:
2-Nitro-1,4-phenylenediamine, 2-Nitrobenzene-1,4-diamine
C12195500
EHERC12195500EA
44
EUR
EHERC12195500
2-Nitro-p-phenylendiamin
2-Nitro-p-phenylendiamin
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,14 g/mol Schmelzpunkt: 134…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007903 CAS-Nummer: 5307-14-2 EINECS: 226-164-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...