2,5-Diaminotoluolsulfat
Lieferant: EHRENSTORFER
C12198500
EHERC12198500EA
38
EUR
EHERC12198500
2,5-Diaminotoluolsulfat
2,5-Diaminotoluolsulfat
Formel:
C₇H₁₂N₂O₄S |
MDL:
MFCD00013003 CAS-Nummer: 615-50-9 EINECS: 210-431-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...