4,4'-Dibromoctafluorbiphenyl (DBOB)

Lieferant: EHRENSTORFER

Synonyme: 4,4-Dibromooctafluorobiphenyl

C12240600
EHERC12240600EA 84 EUR
EHERC12240600
4,4'-Dibromoctafluorbiphenyl (DBOB)
4,4'-Dibromoctafluorbiphenyl (DBOB)
Formel: C₁₂Br₂F₈
Molecular Weight: 455.94 g/mol
MDL: MFCD00000310
CAS-Nummer: 10386-84-2
EINECS: 233-847-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR