Dichlofenthion (O-2,4-Dichlorphenyl-O,O-diethylthiophosphat)

Lieferant: EHRENSTORFER

C12290000
EHERC12290000EA 63 EUR
EHERC12290000
Dichlofenthion (O-2,4-Dichlorphenyl-O,O-diethylthiophosphat)
Dichlofenthion (O-2,4-Dichlorphenyl-O,O-diethylthiophosphat)
Formel: C₁₀H₁₃Cl₂O₃PS
Molecular Weight: 315,16 g/mol
CAS-Nummer: 97-17-6
EINECS: 202-564-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR