3,3'-Dichlorbenzidin

Lieferant: EHRENSTORFER

C12377900
EHERC12377900EA 80 EUR
EHERC12377900
3,3'-Dichlorbenzidin
3,3'-Dichlorbenzidin
Formel: C₁₂H₁₀Cl₂N₂
CAS-Nummer: 91-94-1
EINECS: 202-109-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR