3,3'-Dichlorbenzidin Dihydrochlorid

Lieferant: EHRENSTORFER

C12378000
EHERC12378000EA 63 EUR
EHERC12378000
3,3'-Dichlorbenzidin Dihydrochlorid
3,3'-Dichlorbenzidin Dihydrochlorid
Formel: C₁₂H₁₂Cl₄N₂
CAS-Nummer: 612-83-9
EINECS: 210-323-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR