2,4'-Dichlorbenzophenon
Lieferant: EHRENSTORFER
Synonyme:
2,4'-DBP
C12409000
EHERC12409000EA
85
EUR
EHERC12409000
2,4'-Dichlorbenzophenon
2,4'-Dichlorbenzophenon
Formel:
C₁₃H₈Cl₂O Molecular Weight: 251,11 g/mol |
MDL:
MFCD00038744 CAS-Nummer: 85-29-0 EINECS: 201-596-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...