4,4'-Dichlorbenzophenon
Lieferant: EHRENSTORFER
Synonyme:
4,4-DBP, Bis(4-chlorphenyl)keton
C12410000
EHERC12410000EA
84
EUR
EHERC12410000
4,4'-Dichlorbenzophenon
4,4'-Dichlorbenzophenon
Formel:
C₁₃H₈Cl₂O Molecular Weight: 251,11 g/mol |
MDL:
MFCD00000623 CAS-Nummer: 90-98-2 EINECS: 202-030-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...