2,6-Dichlor-p-kresol
Lieferant: EHRENSTORFER
Synonyme:
1,3-Dichlor-2-hydroxy-5-methylbenzol, 3,5-Dichlor-4-hydroxytoluol, 2,6-Dichlor-4-methylphenol
C12427100
EHERC12427100EA
97
EUR
EHERC12427100
2,6-Dichlor-p-kresol
2,6-Dichlor-p-kresol
Formel:
C₇H₆Cl₂O Molecular Weight: 177,03 g/mol |
MDL:
MFCD00134679 CAS-Nummer: 2432-12-4 EINECS: 219-402-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...