(2,4-Dichlorphenyl)essigsäure
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
DCAA
C12468000
EHERC12468000EA
25.4
EUR
EHERC12468000
(2,4-Dichlorphenyl)essigsäure
(2,4-Dichlorphenyl)essigsäure
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 316 Schmelzpunkt: 132…133 °C |
MDL:
MFCD00004318 CAS-Nummer: 19719-28-9 EINECS: 243-248-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...