Diclofenac

Lieferant: EHRENSTORFER

Synonyme: Diclofenac acid

C12537000
EHERC12537000EA 66 EUR
EHERC12537000
Diclofenac
Diclofenac
Formel: C₁₄H₁₁Cl₂NO₂
CAS-Nummer: 15307-86-5
EINECS: 239-348-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR