4-(Diethylamino)azobenzol
Lieferant: EHRENSTORFER
Synonyme:
N,N-Diethyl-4-(phenyldiazenyl)aniline
C12604600
EHERC12604600EA
85
EUR
EHERC12604600
4-(Diethylamino)azobenzol
4-(Diethylamino)azobenzol
Formel:
C₁₆H₁₉N₃ Molecular Weight: 253,35 g/mol |
MDL:
MFCD00009043 CAS-Nummer: 2481-94-9 EINECS: 219-616-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...